missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bel-Art™ SP Scienceware™ Cleanware™ Self-Adhesive Waste Bags
Disposable waste bags free bench or hood space when hung from walls. Bel Art™ SP Scienceware™ Cleanware™ Self-Adhesive Waste Bags adhere to cabinets and other surfaces.
Manufacturer: Bel-Art™ F131741012
Description
- Peel backing off the adhesive strip across the top and hang close to your work area
- Back panel is longer than the front for easy opening
- High Molecular Weight High Density (HMHD) polyethylene; steam autoclavable at 121°C (250°F)
- Maximum weight of contents should not exceed 1.36kg (3lb)
Specifications
16 x 12 in. (40.64 x 30.48cm) | |
White | |
Cleanware | |
16 in. | |
HDPE (High-density Polyethylene) | |
30.48cm |
1 mil | |
Yes | |
Self-adhesive strip | |
40.64cm | |
12 in. |