missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Cleanware™ Self-Adhesive Waste Bags

Catalog No. p-3273769
Change view
Click to view available options
Dimensions (L x W):
10 x 8 in.
16 x 12 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (L x W)
0181511F 10 x 8 in.
0181511G 16 x 12 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 0181511F Supplier Bel-Art™ Supplier No. F131741008
Only null left

Disposable waste bags free bench or hood space when hung from walls. Bel Art™ Cleanware™ Self-Adhesive Waste Bags adhere to cabinets and other surfaces.

  • Peel backing off the adhesive strip across the top and hang close to your work area
  • Back panel is longer than the front for easy opening
  • High Molecular Weight High Density (HMHD) polyethylene; steam autoclavable at 121°C (250°F)
  • Maximum weight of contents should not exceed 1.36kg (3lb)

Specifications

Autoclavable Autoclavable
Closure Type Zip Tie
Color White
Disposable Single-use
For Use With (Application) Cleanware
For Use With (Equipment) General Trash
Includes Self-adhesive strip
Material HDPE
Sterility Non-sterile
Thickness (English) 1 mil
Dimensions (L x W) 10 x 8 in.
Length (English) 10 in.
Width (English) 8 in.
Product Type Laboratory Waste Bag
Capacity (English) 0.5 gal.
Capacity (Metric) 2 L
Length (Metric) 25.4 cm
Width (Metric) 20.32 cm
Thickness (Metric) 0.025 mm
Show More Show Less