missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Bel-Art™ SP Scienceware™ Clavies™ High-Temperature Autoclave Bags

Withstand high temperatures to reduce autoclave time

Supplier:  Bel-Art™ H131821015

 View more versions of this product

Catalog No. 01-815-3B



Description

Description

Withstand high temperatures to reduce autoclave time

  • Suitable for discarding Petri dishes, membrane filters, multi-well culture plates, pipets and more
  • Rugged, transparent, 0.05mm (2 mil) thick polypropylene
  • Steam autoclavable at 275°F (135°C)

Specifications

Specifications

Autoclavable
Flat
Polypropylene
15 x 10 in. (38.1 x 25.4cm)
38.1 cm
25.4 cm
1.5 gal.
Clear
0.050 mm
15 in.
10 in.
SDS
Documents

Documents

Promotions

Promotions