Learn More
Bel-Art™ Clavies™ High-Temperature Autoclave Bags
Withstand high temperatures to reduce autoclave time.
$84.88
Specifications
Autoclavable Autoclavable
Certifications/Compliance Dart Impact: 1000g per ASTM D1709-01. Tear Resistance: ASTM D1922-00A, MD 200g.
Color Clear
Disposable Single-use
Material Polypropylene
View More Specs
Includes: Bag closures and holders sold separately.
Products 7
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
Catalog Number Mfr. No. Dimensions (L x W) Price Quantity & Availability  
018153A
View Documents Promotion Details
Bel-Art™
H131821010
10 x 10 in.
Pack of 100 Each for $84.88
Only null left
018153
View Documents Promotion Details
Bel-Art™
H131821012
12 x 10 in. (30.48 x 25.4cm)
Pack of 100 Each for $84.88
Only null left
018153B
View Documents Promotion Details
Bel-Art™
H13182 1015
15 x 10 in. (38.1 x 25.4cm)
Pack of 100 Each for $84.88
Only null left
018154
View Documents Promotion Details
Bel-Art™
H131852436
36 x 24 in. (91 x 61 cm)
Pack of 100 Each for $84.88
Only null left
018155
View Documents Promotion Details
Bel-Art™
H13185 2430
30 x 24 in. (76 x 61 cm)
Pack of 100 Each for $84.88
Case of 10 Pack for $7,021.40
Only null left
018157
View Documents Promotion Details
Bel-Art™
H13185 1224
24 x 12 in. (61 x 30 cm)
Pack of 100 Each for $84.88
Case of 10 Pack for $2,669.00
Only null left
018156
View Documents Promotion Details
Bel-Art™
H131820812
12 x 8 in. (30 x 20 cm)
Pack of 100 Each for $84.88
Case of 10 Pack for $2,131.40
Only null left
Description
Description
Withstand high temperatures to reduce autoclave time
  • Suitable for discarding Petri dishes, membrane filters, multi-well culture plates, pipets and more
  • Rugged, transparent, 0.05 mm (2 mil) thick polypropylene
  • Steam autoclavable at 275°F (135°C)
Specifications
Specifications
Autoclavable
Clear
Polypropylene
Autoclave Bags
Dart Impact: 1000g per ASTM D1709-01. Tear Resistance: ASTM D1922-00A, MD 200g.
Single-use
2 mil