missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Clavies™ Biohazard Bag Holders
Keep bags open and ready to use
$140.31 - $419.64
Specifications
| Autoclavable | Autoclavable |
|---|---|
| Color | Silver, Orange |
| Disposable | Reusable |
| For Use With (Application) | Designed to securely support the bag and its contents |
| Material | Stainless Steel |
| Catalog Number | Mfr. No. | Height (English) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Height (English) | Price | Quantity & Availability | |||||
|
0181612
|
Bel-Art™
F131920003 |
27.2 in. |
Each for $419.64
|
|
|||||
|
0181610
|
Bel-Art™
F131920001 |
18.5 in. |
Each for $140.31
|
|
|||||
|
0181611
|
Bel-Art™
F131920002 |
21.5 in. |
Each for $140.31
|
|
|||||
Description
- Economical, lightweight stainless-steel wire frame
- Highly visible orange polypropylene bottom tray for easy identification as a biohazard containment system
- Tray serves as containment well in case of bag leakage
- Autoclavable at 121°C (250°F) and dishwasher safe
- Assembly is easy and permanent
- Medium and large models can be used with autoclavable polypropylene cover(Mfr. No. F13192-0102) to improve sanitation
Specifications
| Autoclavable | |
| Reusable | |
| Stainless Steel | |
| Benchtop |
| Silver, Orange | |
| Designed to securely support the bag and its contents | |
| Sterile | |
| Biohazard Bag Holder |