missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bel-Art™ Clavies™ Biohazard Bag Holders
Keep bags open and ready to use
Supplier: Bel-Art™ F131920002
Description
- Economical, lightweight stainless-steel wire frame
- Highly visible orange polypropylene bottom tray for easy identification as a biohazard containment system
- Tray serves as containment well in case of bag leakage
- Autoclavable at 121°C (250°F) and dishwasher safe
- Assembly is easy and permanent
- Medium and large models can be used with autoclavable polypropylene cover(Mfr. No. F13192-0102) to improve sanitation
Chemical Identifiers
| Autoclavable | |
| Reusable | |
| Stainless Steel | |
| Benchtop |
| Silver, Orange | |
| Designed to securely support the bag and its contents | |
| Sterile | |
| Biohazard Bag Holder |
Specifications
| Autoclavable | |
| Reusable | |
| 01-826B, 01-826-6, 01-827C, 01-814-2, 01-815B medium 61 x 76cm Clavies™ biohazard bag | |
| Stainless Steel | |
| Benchtop | |
| 24 in. | |
| 54.6 cm | |
| 61 cm |
| Silver, Orange | |
| Designed to securely support the bag and its contents | |
| Stainless steel wire frame and orange polypropylene bottom, plastic wire anchors, wire locking clips | |
| Sterile | |
| Medium | |
| 21.5 in. | |
| Biohazard Bag Holder |