missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ Automatic Self-Zeroing Glass Burets

Closed system prevents outside contamination

Supplier:  Bel-Art™ F379151010

 View more versions of this product

Catalog No. 03-831


Get More Bel-Art Lab Products with Purchase
View all promotions
Only null left

Includes: Drain arm with 2mm bore glass stopcock.

Description

Description

  • Squeeze the flexible polyethylene bottle to force liquid into the buret, overfill automatically drains back into reservoir
  • Self-zeroing borosilicate glass burets have easy to view white 0.1 mL graduations
  • Glass stopcock on drain arm has 2 mm bore for smooth, controlled dispensing
  • Large, stable plastic base can be detached from reservoir for cleaning
  • Class A graduation accuracy, within 1% of total volume
Specifications

Specifications

+/- 1%
Screw Cap
10 mL
18.5 in.
0.1 mL
47 cm
White
Glass
None
Automatic Buret
Not Autoclavable
Natural
47 cm
Borosilicate Glass
Titration, Reagent Dispensing
Polyethylene Bottle, Stable Plastic Base
Glass
Class A
1000 mL
Class A
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions