missing translation for 'onlineSavingsMsg'
Learn More

Behenamide 75.0+%, TCI America™

Catalog No. p-7126551
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D100725G 25 g
D1007500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D100725G Supplier TCI America Supplier No. D100725G
Only null left

Chemical Identifiers

CAS 3061-75-4
Molecular Formula C22H45NO
Molecular Weight (g/mol) 339.608
MDL Number MFCD00047905
InChI Key ORAWFNKFUWGRJG-UHFFFAOYSA-N
Synonym behenamide, behenic acid amide, behenic amide, docosanamid, behenylamide, docsoamide, kemamide b, uniwax 1747, acmc-209hhe, behenic amide, tech
PubChem CID 76468
IUPAC Name docosanamide
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)N

Specifications

Melting Point 111°C
Color White-Yellow
Quantity 25 g
Formula Weight 339.61
Percent Purity ≥75.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Behenamide
EINECSNumber (2)-0824
TSCA Yes