accessibility menu, dialog, popup

UserName

CAS RN 3061-75-4

H3CNH2O

CAS RN 3061-75-4

IUPAC Name: docosanamide
Synonyms: behenamide
Molecular Weight (g/mol): 339.61
Molecular Formula: C22H45NO
InChi Key: ORAWFNKFUWGRJG-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCCCCC(N)=O
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results