missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Behenamide 75.0+%, TCI America™
Specifications
Specifications
| Melting Point | 111°C |
| Color | White-Yellow |
| Quantity | 500 g |
| Formula Weight | 339.61 |
| Percent Purity | ≥75.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Behenamide |
Safety and Handling
| EINECSNumber | (2)-0824 |
| TSCA | Yes |