missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BEGAIN Monoclonal Antibody (OTI4D9)

Description
BEGAIN Monoclonal Antibody for Western Blot
May sustain the structure of the postsynaptic density (PSD). Target protein binds the guanylate kinase domain of PSD-95/SAP90, may play a role in the post-synaptic membrane.
Specifications
Specifications
| Antigen | BEGAIN |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | OTI4D9 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | BEGAIN |
| Gene Accession No. | Q9BUH8 |
| Gene Alias | BEGAIN; BM948371; brain enriched guanylate kinase associated; brain-enriched guanylate kinase-associated; brain-enriched guanylate kinase-associated homolog; brain-enriched guanylate kinase-associated protein; brain-enriched guanylate kinase-associated; brain-enriched guanylate kinase-associated protein; Gm897; KIAA1446 |
| Show More |