Learn More
Supertek Beaker with Handle (Measuring Jug)
Polypropylene Beaker with Handle (Measuring Jug) that is autoclavable and has a non-drip spout. These jugs have raised graduations with contact clarity, and are provided with a thumb grip on the handle.
Supplier: Supertek CH101659
Description
Sturdy polypropylene construction makes these beakers durable, with excellent chemical resistance.
- Polypropylene
- Autoclavable
- Raised graduations
- Large, strong handles with thumb grip for safe handling of acids, alkalis, and other hazardous chemicals.
Specifications
| 5000 mL | |
| Beaker | |
| Reusable | |
| Embossed Scale |
| Polypropylene | |
| Autoclavable | |
| Yes | |
| Handled |