missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Supertek Beaker with Handle (Measuring Jug)
Polypropylene Beaker with Handle (Measuring Jug) that is autoclavable and has a non-drip spout. These jugs have raised graduations with contact clarity, and are provided with a thumb grip on the handle.
$48.57 - $145.70
Specifications
| Material | Polypropylene |
|---|---|
| Type | Beaker |
| Autoclavable | Autoclavable |
| Disposable | Reusable |
| Graduated | Yes |
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Capacity (Metric) | Price | Quantity | |||||
S01877
|
Supertek
CH101631SP |
250 mL |
N/A
|
|
|||||
S01878
|
Supertek
CH101651 |
500 mL |
N/A
|
|
|||||
S01875
|
Supertek
CH101653 |
1000 mL |
N/A
|
|
|||||
S01876
|
Supertek
CH101655 |
2000 mL |
N/A
|
|
|||||
S01873
|
Supertek
CH101657 |
3000 mL |
N/A
|
|
|||||
S01874
|
Supertek
CH101659 |
5000 mL |
N/A
|
|
|||||
Description
Sturdy polypropylene construction makes these beakers durable, with excellent chemical resistance.
- Polypropylene
- Autoclavable
- Raised graduations
- Large, strong handles with thumb grip for safe handling of acids, alkalis, and other hazardous chemicals.
Specifications
| Polypropylene | |
| Autoclavable | |
| Yes | |
| Handled |
| Beaker | |
| Reusable | |
| Embossed Scale |