missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Rat BDNF ELISA Kit

ELISA

$936.00

Specifications

Accession Number P23363
Assay Range 12.29 to 3000 pg/mL
Assay Sensitivity 12 pg/mL
Calibration Range 12 pg/mL to 3,000 pg/mL
Description BDNF ELISA Kit, 96 Assays, 1 Kit, Rat Species, HRP Conjugate, Colorimetric Detection Method, Enzyme-Linked Immunosorbent Assay Designed for the Quantitation and Detection of BDNF in Samples Using a Microplate Reader
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ERBDNF
Encompass
View Documents Promotion Details
Invitrogen™
ERBDNF
96 Tests
Pack for $936.00
Only null left
 
Description

Description

The Rat Brain-Derived Neurotrophic Factor (Rt BDNF) ELISA quantitates Rt BDNF in rat serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Rt BDNF. Principle of the method The Rat BDNF solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

BDNF (Brain-Derived Neurotrophic Factor) is a member of the neurotrophin family. BDNF is synthesized as pre-proBDNF, followed by cleavage to proBDNF. Although further processing generates the mature, 14 kDa protein, pro-BDNF is biologically active and is secreted in synaptic vesicles along with the mature form. BDNF is widely expressed in the central nervous system and acts in an autocrine and paracrine manner on several classes of neurons. Signaling occurs mainly through the tyrosine kinase receptor TrkB, although binding to the lower-affinity receptor, p75-NTR, has also been demonstrated. BDNF promotes neuronal survival and differentiation and has been shown to play a critical role in memory formation and synaptic regulation. BDNF is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of BDNF is reduced in both Alzheimer's and Huntington disease patients, and may also play a role in the regulation of stress response and in the biology of mood disorders.
Specifications

Specifications

P23363
12 pg/mL
BDNF ELISA Kit, 96 Assays, 1 Kit, Rat Species, HRP Conjugate, Colorimetric Detection Method, Enzyme-Linked Immunosorbent Assay Designed for the Quantitation and Detection of BDNF in Samples Using a Microplate Reader
Solid-phase sandwich ELISA
Colorimetric
Growth Factors & Receptors
This ELISA pair antibody detects rat and human BDNF. Other species not determined.
ELISA, Protein Assays, Protein Biology, Cancer Biology, Immunology, Neuroscience, Neurobiology, Protein Detection, Protein Analysis
24225
Rat
<10
HRP
Neurobiology
2°C to 8°C
1 hrs 20 mins
12.29 to 3000 pg/mL
12 pg/mL to 3,000 pg/mL
Rat and human BDNF
HRP
ELISA
Plasma, Serum, Supernatant
96 Assays
Colorimetric Microplate Reader
Bdnf
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
Plasma 10 μL, Serum 10 μL, Supernatant 100 μL
BDNF
4 hrs 45 mins
Videos
SDS
Documents

Documents