missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BDNF Monoclonal Antibody (C4)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA565974
Description
BDNF Monoclonal Antibody for ICC/IF, IHC (P), Western Blot
BDNF (Brain-Derived Neurotrophic Factor) is a member of the neurotrophin family. BDNF is synthesized as pre-proBDNF, followed by cleavage to proBDNF. Although further processing generates the mature, 14 kDa protein, pro-BDNF is biologically active and is secreted in synaptic vesicles along with the mature form. BDNF is widely expressed in the central nervous system and acts in an autocrine and paracrine manner on several classes of neurons. Signaling occurs mainly through the tyrosine kinase receptor TrkB, although binding to the lower-affinity receptor, p75-NTR, has also been demonstrated. BDNF promotes neuronal survival and differentiation and has been shown to play a critical role in memory formation and synaptic regulation. BDNF is induced by cortical neurons, and is necessary for survival of striatal neurons in the brain. Expression of BDNF is reduced in both Alzheimer's and Huntington disease patients, and may also play a role in the regulation of stress response and in the biology of mood disorders.Specifications
| BDNF | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| P14082, P21237, P23363, P23560, P25429, Q0EAB7 | |
| BDNF | |
| Recombinant Brain Derived Neurotrophic Factor (BDNF) (Pro20-Arg252). | |
| 200 μL | |
| Primary | |
| Human, Mouse, Rat, Rabbit, Pig, Horse, Chicken, Goat | |
| Antibody | |
| IgG2b κ |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| C4 | |
| Unconjugated | |
| BDNF | |
| abrineurin; ANON2; anorexia BDNF; Bdnf; BDNF precursor form; bdnf protein; brain derived neurothrophic factor; brain derived neurotrophic factor; brain-derived neurotrophic factor; BULN2; H-BDNF; neurotrophin; ProBDNF | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 100009689, 100356949, 102180782, 12064, 24225, 396186, 397495, 627 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |