missing translation for 'onlineSavingsMsg'
Learn More

BDE No 47 Solution, 50 μg/mL in isooctane, MilliporeSigma™ Supelco™

Catalog No. 111018129
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018129 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018129 Supplier Merck Emd Millipore Supplier No. 336701ML
Only null left
Environmental

Specifications

CAS 5436-43-1
Flash Point -12°C
Linear Formula C12H6 Br4 O
MDL Number MFCD00626457
Synonym 2,2 ′,4,4 ′-Tetrabromodiphenyl ether solution; 2,2 ′,4,4 ′-TetraBDE; PBDE 47
Type Analytical Standard
Quantity 1 mL
Formula Weight 485.79
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 50 μg/mL in isooctane