missing translation for 'onlineSavingsMsg'
Learn More

BDE No 47, ≥97% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014554
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014554 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014554 Supplier Merck Emd Millipore Supplier No. 9183410MG
Only null left

BDE-47 belongs to the class of polybrominated diphenyl ethers (PBDEs), commonly used as fire retardants in consumer products.

Specifications

CAS 5436-43-1
Linear Formula C12H6 Br4 O
MDL Number MFCD00626457
Synonym 2,2′,4,4′-TetraBDE; 2,2′,4,4′-Tetrabromodiphenyl ether; PBDE 47
Type Analytical Standard
Quantity 10 mg
Formula Weight 485.79
Percent Purity ≥97% (GC)
Grade Analytical Standard