missing translation for 'onlineSavingsMsg'
Learn More

BDE No 28 Solution, 50 μg/mL in isooctane, MilliporeSigma™ Supelco™

Catalog No. 111018128
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018128 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018128 Supplier Merck Emd Millipore Supplier No. 336631ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BDE No 28 is a ubiquitous environmental contaminant with low acute toxicity, belonging to the class of polybrominated diphenyl ethers. It can find wide applications as a flame-retardant additive in consumer products ranging from textiles to polymers.

Environmental

Specifications

CAS 41318-75-6
Flash Point -12°C
Linear Formula C12H7 Br3 O
MDL Number MFCD09037597
Synonym 2,4,4 ′-Tribromodiphenyl ether solution; 2,4,4 ′-TriBDE; PBDE 28
Type Analytical Standard
Quantity 1 mL
Formula Weight 406.90
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 50 μg/mL in isooctane