missing translation for 'onlineSavingsMsg'
Learn More

BDE No 28, ≥97% (GC), MilliporeSigma™ Supelco™

Catalog No. 111014160
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014160 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014160 Supplier Merck Emd Millipore Supplier No. 0536410MG
Only null left

Specifications

CAS 41318-75-6
Linear Formula C12H7 Br3 O
MDL Number MFCD09037597
Melting Point 63°C to 66°C
Synonym 2,4,4′-TriBDE; 2,4,4′-Tribromodiphenyl ether; PBDE 28
Type Analytical Standard
Quantity 10 mg
Formula Weight 406.90
Percent Purity ≥97% (GC)
Grade Analytical Standard