missing translation for 'onlineSavingsMsg'
Learn More

BDE No 183 Solution, 50 μg/mL in isooctane, MilliporeSigma™ Supelco™

Catalog No. 111018101
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111018101 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111018101 Supplier Merck Emd Millipore Supplier No. 336861ML

BDE No 183 belongs to the class of polybrominated diphenyl ethers. It can be commercially used as a flame retardant additive in building materials, textiles, plastics and electronic equipment.

Environmental

Specifications

CAS 207122-16-5
Flash Point -12°C
Linear Formula C12H3 Br7 O
MDL Number MFCD16661229
Synonym 2,2 ′,3,4,4 ′,5 ′,6-Heptabromodiphenyl ether solution; 2,2 ′,3,4,4 ′,5 ′,6-HeptaBDE; PBDE 183
Type Analytical Standard
Quantity 1 mL
Formula Weight 722.48
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 50 μg/mL in isooctane