missing translation for 'onlineSavingsMsg'
Learn More

BD140 98.0+%, TCI America™

Catalog No. D489825MG
Change view
Click to view available options
Quantity:
25 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D489825MG 25 mg
D4898100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D489825MG Supplier TCI America Supplier No. D489825MG

Chemical Identifiers

CAS 1201643-08-4
Molecular Formula C21H21BF2N2O
Molecular Weight (g/mol) 366.219
InChI Key NDQRWYKJLFZGBK-RMKNXTFCSA-N
Synonym 4,4-Difluoro-1-methyl-3-(4-propoxystyryl)-4-bora-3a,4a-diaza-s-indacene
PubChem CID 72198885
SMILES [B-]1(N2C(=CC(=C2C=C3[N+]1=CC=C3)C)C=CC4=CC=C(C=C4)OCCC)(F)F

Specifications

Melting Point 155°C
Quantity 25 mg
Formula Weight 366.22
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material BD140 [for Albumin binding assay]