missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Vacutainer™ Urine Collection Kits: Urinalysis
Reduce the need to retest, relabel, and/or recollect specimens
Supplier: BD 364991
Description
- Closed systems that benefit healthcare workers by reducing their need to come into contact with potentially hazardous specimens
- Designed to provide safety and satisfaction, as well as consistent, reliable results to ensure proper patient outcomes
364991 Urinalysis Transfer Straw Kit: 8 ml draw Plus plastic conical bottom tube. The BD Vacutainer urine collection straw is a transfer device that lets you transfer urine to one or more tubes without being exposed to the specimen.
Chemical Identifiers
| Plastic | |
| Sterile | |
| For UA chemistry Urine testing | |
| Room temperature |
Specifications
| Urine Collection Kit | |
| Sterile | |
| For UA chemistry Urine testing | |
| Yes | |
| 0.27 oz. |
| Plastic | |
| Conical | |
| Transfer Device, Evacuated Tube | |
| Room temperature | |
| 8 mL |