missing translation for 'onlineSavingsMsg'
Learn More

BD Vacutainer™ Urine Collection Kits: Urinalysis

Catalog No. p-4901676
Change view
Click to view available options
Volume (English):
0.27 oz.
0.33 oz.
Volume (Metric):
8 mL
10 mL
Includes:
Evacuated Conical Bottom Tube
Evacuated Round Bottom Tube
Evacuated Tube, Cup
Preservative, Transfer Device, Tube
Preservative, Transfer Device, Tube, Cup
Transfer Device, Evacuated Tube
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (English) Volume (Metric) Includes
1482793 0.27 oz. 8 mL Preservative, Transfer Device, Tube, Cup
1482791 0.27 oz. 8 mL Preservative, Transfer Device, Tube
02683123 0.33 oz. 10 mL Evacuated Tube, Cup
02683126 0.27 oz. 8 mL Evacuated Tube, Cup
02683128 0.27 oz. 8 mL Transfer Device, Evacuated Tube
02683100 0.27 oz. 8 mL Evacuated Conical Bottom Tube
14375138 0.33 oz. 10 mL Evacuated Round Bottom Tube
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 1482793 Supplier BD Supplier No. 364946
Only null left

Reduce the need to retest, relabel, and/or recollect specimens

  • Closed systems that benefit healthcare workers by reducing their need to come into contact with potentially hazardous specimens
  • Designed to provide safety and satisfaction, as well as consistent, reliable results to ensure proper patient outcomes

Urinalysis Products

  • Include BD™ proprietary mercury-free Urinalysis Preservative Tube, which provides compliance with both the Memorandum of Understanding for the U.S. EPA and the AHA
  • Help to maintain urine sample integrity for up to 72 hours at room temperature
  • Minimum and maximum fill lines (7-8mL) ensure proper additive-to-urine ratio

Specifications

Product Type Urine Collection Kit
Material Plastic
Sterility Sterile
Bottom Shape Conical
For Use With (Application) For UA chemistry Urine testing
Includes Preservative, Transfer Device, Tube, Cup
Latex Free Yes
Storage Requirements Room temperature
Volume (English) 0.27 oz.
Volume (Metric) 8 mL