missing translation for 'onlineSavingsMsg'
Learn More

BD GasPak™ EZ Gas Generating Systems and Supplies: Container System Sachets

Used in producing anaerobic, microaerophilic, or CO2-enriched environments. BD Difco™ GasPak™ EZ Gas Generating Systems and Supplies: Container System Sachets provide catalyst-free convenience.

$201.50 - $252.26

Specifications

Content And Storage 2°C to 25°C
Format Sachet
For Use With (Application) Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
Product Line GasPak
View More Specs

Products 4
Catalog Number Mfr. No. Type For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. Type For Use With (Equipment) Price Quantity & Availability  
B260001
View Documents Promotion Details
BD
260001
Sachet w/ Indicator EZ Anaerobe Container System
Pack of 20 Each for $252.26
Only null left
 
B260678
View Documents Promotion Details
BD
260678
Sachet EZ Anaerobe Container System
Pack of 20 Each for $240.21
Only null left
 
B260680
View Documents Promotion Details
BD
260680
Sachet EZ Campy Container System
Pack of 20 Each for $245.97
Only null left
 
B260679
View Documents Promotion Details
BD
260679
Sachet EZ CO2 Container System
Pack of 20 Each for $201.50
Only null left
 
Description

Description

  • For use in both the GasPak™ EZ Containers and the GasPak™ 100 and GasPak™ 150 jars
  • Sachets require no water or catalyst for activation and are engineered to provide rapid gas generation for optimal bacterial growth
Specifications

Specifications

2°C to 25°C
Sachet
Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
GasPak
Videos
SDS
Documents

Documents

Catalyst-free convenience for use in producing anaerobic, microaerophilic, or CO2-enriched environments