missing translation for 'onlineSavingsMsg'
Learn More

BD GasPak™ EZ Gas Generating Systems and Supplies: Container System Sachets

Catalog No. B260679
Change view
Click to view available options
Type:
Sachet
Sachet w/ Indicator
For Use With (Equipment):
EZ Anaerobe Container System
EZ Campy Container System
EZ CO2 Container System
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type For Use With (Equipment)
B260679 Sachet EZ CO2 Container System
B260001 Sachet w/ Indicator EZ Anaerobe Container System
B260678 Sachet EZ Anaerobe Container System
B260680 Sachet EZ Campy Container System
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. B260679 Supplier BD Supplier No. 260679
Only null left

Used in producing anaerobic, microaerophilic, or CO2-enriched environments. BD Difco™ GasPak™ EZ Gas Generating Systems and Supplies: Container System Sachets provide catalyst-free convenience.

  • For use in both the GasPak™ EZ Containers and the GasPak™ 100 and GasPak™ 150 jars
  • Sachets require no water or catalyst for activation and are engineered to provide rapid gas generation for optimal bacterial growth

Specifications

Type Sachet
Content And Storage 2°C to 25°C
Format Sachet
For Use With (Application) Primary isolation and cultivation of anaerobic, microaerophilic or capnophilic bacteria by use of gas generating sachets
For Use With (Equipment) EZ CO2 Container System
Product Line GasPak
Quantity 20 Sachets/Box

Catalyst-free convenience for use in producing anaerobic, microaerophilic, or CO2-enriched environments