Learn More
BD Difco™ Neisseria Meningitidis Antisera
For serotyping Neisseria meningitidis in slide agglutination tests
$624.13
Specifications
| Certifications/Compliance | For in vitro diagnostic use |
|---|---|
| Product Type | Antiserum |
Alert:
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Description
- Stable, lyophilized, polyclonal rabbit antisera
- Contains 0.1% sodium azide as preservative
Specifications
| For in vitro diagnostic use | |
| Antiserum |
For in vitro diagnostic use.