missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Neisseria Meningitidis Antisera

Catalog No. DF2252500
Change view
Click to view available options
Quantity:
1 mL
Description:
Antiserum Group Z prime (Z' )
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Description
DF2252500 1 mL Antiserum Group Z prime (Z' )
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.
Catalog No. DF2252500 Supplier BD Supplier No. 222521

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The products indicated contain mercury. Don't put products containing mercury in the trash. Products containing mercury are to be recycled or disposed of as hazardous waste. Products containing mercury may be regulated by Local, State, or Federal Laws. Some products containing mercury are not available for sale in some states.

For serotyping Neisseria meningitidis in slide agglutination tests

  • Stable, lyophilized, polyclonal rabbit antisera
  • Contains 0.1% sodium azide as preservative

Specifications

Certifications/Compliance For in vitro diagnostic use
Description Antiserum Group Z prime (Z' )
Product Type Neisseria Meningitidis Antisera
Packaging Type Bottle
Quantity 1 mL

For in vitro diagnostic use.