missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Agar, Technical

Catalog No. p-4886727
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Container
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging Type
DF0812071 2 kg Container
DF0812179 500 g Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. DF0812071 Supplier BD Supplier No. 281210
Only null left

Used in preparing microbiological culture media. BD™ Difco™ Agar, Technical is a solidifying agent with wider quality control parameters than other bacteriological agars. However, the solubility, gelation temperature and solidity are carefully monitored to permit its use.

Technical Agar is suitable for many bacteriological applications. This agar is not highly processed, has broader technical specifications than other agars and is not recommended for growth of fastidious organisms. It shows good cultural response (in nutrient broth) to Escherichia coli 25922 and Staphylococcus aureus 25923.

  • Loss on Drying (LOD): ≤20%
  • Ash(%): ≤6.5%
  • Calcium μg/g (ppm): ≤3,000 ppm
  • Magnesium μg/g (ppm): ≤1,300 ppm
  • Melting Point: ≤85°C
  • Gelation Point: 32–39°C
  • Gel Strength: 613g/cm3
  • pH (1.5% solution, 25°C): 5.9
  • Spore count: 4,300 CFU/g
  • Standard plate count: 2,725 CFU/g
  • Inorganics: Calcium 0.110%, Chloride 0.172%, Cobalt <0.001%, Copper <0.001%, Iron 0.002%, Lead <0.001%, Magnesium 0.093%, Manganese <0.001%, Nitrate <0.005%, Phosphate 0.015%, Potassium 0.124%, Sodium 0.932%, Sulfate 0.367%, Sulfur 0.646%, Tin <0.001%, Zinc <0.001%

Agar is a phycocolloid extracted from a group of red-purple marine algae (Class Rhodophyceae). Agar is the gelling agent of choice because it remains firm at growth temperatures for many pathogens. It is also resistant to a breakdown by bacterial enzymes. Agar melts at a different temperature than that at which it solidifies, a property known as hysteresis. It is generally resistant to shear forces; however, different agars may have different gel strengths. Agar is typically used in a final concentration of 1–2% for solidifying culture media.

Specifications

Certifications/Compliance ISO/DIS, 11014
Color Very light to medium beige
Concentration 1.5%
Content And Storage <30°C
Additive Technical Grade Agar
Form Dehydrated Powder
Grade Technical
For Use With (Application) Bacteriological culture media
Packaging Type Container
Quantity 2 kg