Learn More
Gibco™ Difco™ TC Yeastolate, ultra-filtered (UF)
Description
Difco TC Yeastolate UF Low Bioburden is an animal origin-free and water-soluble extract of autolyzed yeast, enhanced for tissue culture (TC) applications. This product is a mixture of low molecular weight peptides, amino acids, carbohydrates (simple and complex), and vitamins. In addition to the performance-enhancing features of our TC yeastolate product, this offering can help reduce endotoxin and bioburden levels.
Difco TC Yeastolate, ultra-filtered (UF), is an animal origin-free (AOF) product derived from water-soluble portions of autolyzed yeast (Saccharomyces cerevisiae). This product is intended as a nutritional supplement for bacterial, insect, and mammalian cell culture.
Low bioburden
• Reducing bioburden levels limits the effects of potential interactions between the organisms and the peptone, which could impact product stability and performance
• May eliminate the need to remove higher levels of bioburden from incoming critical raw materials, therefore helping to improve the upstream bioproduction process
• Available in bioburden levels of ≤100–300 CFU
Specifications
Specifications
| Color | Beige |
| Content And Storage | Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers. |
| Culture Type | Mammalian, Insect, Microbial |
| Form | Powder |
| Product Type | TC Yeastolate |
| Endotoxin Level | 500 EU/g |
| Format | Pail |
| For Use With (Application) | Preparing bacterial, insect, and mammalian cell culture media |
| Manufacturing Quality | UF |
| pH | 5.4 to 7.2 |
| Show More |