missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Difco™ TC Yeastolate, ultra-filtered (UF)
GREENER_CHOICE

Catalog No. B92804
Change view
Click to view available options
Quantity:
500 g
10 kg
50 kg
Manufacturing Quality:
UF
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Manufacturing Quality
B92804 500 g UF
B92805 10 kg UF
670079 50 kg UF
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B92804 Supplier Gibco™ Supplier No. 292804

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Enhance cell culture performance with Gibco™ Difco™ TC Yeastolate Ultra-Filtered (UF) Low Bioburden while also mitigating risk in bioprocessing methods.

Difco TC Yeastolate UF Low Bioburden is an animal origin-free and water-soluble extract of autolyzed yeast, enhanced for tissue culture (TC) applications. This product is a mixture of low molecular weight peptides, amino acids, carbohydrates (simple and complex), and vitamins. In addition to the performance-enhancing features of our TC yeastolate product, this offering can help reduce endotoxin and bioburden levels.

Difco TC Yeastolate, ultra-filtered (UF), is an animal origin-free (AOF) product derived from water-soluble portions of autolyzed yeast (Saccharomyces cerevisiae). This product is intended as a nutritional supplement for bacterial, insect, and mammalian cell culture.

Low bioburden
• Reducing bioburden levels limits the effects of potential interactions between the organisms and the peptone, which could impact product stability and performance
• May eliminate the need to remove higher levels of bioburden from incoming critical raw materials, therefore helping to improve the upstream bioproduction process
• Available in bioburden levels of ≤100–300 CFU

Specifications

Color Beige
Content And Storage Storage conditions: 2°C to 25°C. Store in cool, dry conditions in well-sealed containers.
Culture Type Mammalian, Insect, Microbial
Form Powder
Product Type TC Yeastolate
Endotoxin Level 500 EU/g
Format Bottle
For Use With (Application) Preparing bacterial, insect, and mammalian cell culture media
Manufacturing Quality UF
pH 5.4 to 7.2
Product Line Difco
Quantity 500 g
Show More Show Less