missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: TAT Broth Base
For sterility testing of viscous materials, adapted to sterility of cosmetics
$408.16 - $1713.87
Specifications
| Content And Storage | <30°C |
|---|---|
| Form | Dehydrated |
| Media Type | TAT Broth Base |
| Product Type | Microbiology Media |
| For Use With (Application) | Microbial growth |
| Catalog Number | Mfr. No. | Quantity | Packaging Type | For Use With (Application) | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging Type | For Use With (Application) | Price | Quantity | |||
|
DF0984-17-1
|
BD
298410 |
500 g | Bottle | Microbial growth |
Each for $408.16
|
|
|||
|
14-910-97
|
BD
292848 |
2000 g | Pail | Microbial growth |
Each for $1,713.87
|
|
|||
Description
For sterility testing of viscous materials, adapted to sterility of cosmetics
Certifications
ISO/DIS, 11014
Specifications
| <30°C | |
| TAT Broth Base | |
| Microbial growth |
| Dehydrated | |
| Microbiology Media |