missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: TAT Broth Base
For sterility testing of viscous materials, adapted to sterility of cosmetics
Supplier: BD 292848
Description
For sterility testing of viscous materials, adapted to sterility of cosmetics
Specifications
| ISO/DIS, 11014 | |
| Dehydrated | |
| Microbiology Media | |
| Microbial growth | |
| 2000 g |
| <30°C | |
| TAT Broth Base | |
| Pancreatic digest of Casein 20g/L, Soy Lecithin 5g/L | |
| Pail |