missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Modified Letheen Agar, (AOAC)
Microbiological evaluation of cosmetics - neutralizes quaternary ammonium compounds
$417.58 - $1321.00
Specifications
| Form | Dehydrated |
|---|---|
| Media Type | Letheen Agar |
| Product Type | Agar |
| For Use With (Application) | Microbiological evaluation of cosmetics - neutralizes quaternary Ammonium compounds |
| Catalog Number | Mfr. No. | Quantity | Packaging Type | Media Type | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging Type | Media Type | Price | Quantity | |||
|
DF0631-17-0
|
BD
263110 |
500 g | Bottle | Letheen Agar |
Each for $417.58
|
|
|||
|
14-910-96
|
BD
292847 |
2 kg | Pail | Letheen Agar |
Each for $1,321.00
|
|
|||
Description
Applications:
For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria
Specifications
| Dehydrated | |
| Letheen Agar | |
| Agar | |
| Microbiological evaluation of cosmetics - neutralizes quaternary Ammonium compounds |