Learn More
BD Difco™ Dehydrated Culture Media: Modified Letheen Agar, (AOAC)
Microbiological evaluation of cosmetics - neutralizes quaternary ammonium compounds
Supplier:  BD 263110
 View more versions of this product
Catalog No. DF0631-17-0
Only null left
Explore available promotions
Description
Description
Applications:
For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria
Specifications
Specifications
BAM
Dehydrated
Agar
Microbiological evaluation of cosmetics - neutralizes quaternary Ammonium compounds
500 g
2°C to 8°C
Letheen Agar
Letheen Agar 32g/L, Tryptone 5g/L, Proteose Peptone No. 3 10g/L, Yeast extract 2g/L, Sodium Chloride 5g/L, Sodium Bisulfite 0.1g/L, Agar 5g/L
Bottle
Product Suggestions
Product Suggestions