missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Luria Broth Base, Miller
For propagation and maintenance of E. coli for use in molecular biology
$355.76 - $1187.66
Specifications
| Certifications/Compliance | CE, COMPF |
|---|---|
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Luria Broth Base, Miller |
| Product Type | Microbiology Media |
| Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging Type | Price | Quantity | ||||
|
DF0414-07-3
|
BD
241410 |
2 kg | Pail |
Each for $1,187.66
|
|
||||
|
DF0414-17-1
|
BD
241420 |
500 g | Bottle |
Each for $355.76
|
|
||||
Description
- Contains 0.5g/L NaCl
Applications:
For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria
Specifications
| CE, COMPF | |
| Dehydrated | |
| Microbiology Media | |
| Maintaining and propagating Escherichia coli in molecular microbiology procedures with or without added Glucose |
| Refrigerate | |
| Luria Broth Base, Miller | |
| Pancreatic digest of Casein 10g/L, Yeast extract 5g/L, Sodium Chloride 0.5g/L |