missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Luria Broth Base, Miller

For propagation and maintenance of E. coli for use in molecular biology

Supplier:  BD 241410

 View more versions of this product

Catalog No. DF0414-07-3


Only null left
Explore available promotions

Description

Description

  • Contains 0.5g/L NaCl

Applications:

For evaluation, maintenance, selective enrichment, differentiation, isolation, cultivation, detection, propagation of bacteria

Specifications

Specifications

CE, COMPF
Dehydrated
Microbiology Media
Maintaining and propagating Escherichia coli in molecular microbiology procedures with or without added Glucose
2 kg
Refrigerate
Luria Broth Base, Miller
Pancreatic digest of Casein 10g/L, Yeast extract 5g/L, Sodium Chloride 0.5g/L
Pail
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions