missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Lauryl Tryptose Broth
Medium for detection of coliform bacteria
Supplier: BD 224120
Specifications
| AOAC, BAM, CCAM, COMPF, ISO, SMD, SMWW | |
| Dehydrated | |
| Microbiology Media | |
| Detection of Coliform organisms in materials of sanitary importance | |
| 2 kg |
| Refrigerate | |
| Lauryl Tryptose Broth | |
| Tryptose 20g/L, Lactose 5g/L, Dipotassium Phosphate 2.75g/L, Monopotassium Phosphate 2.75g/L, Sodium Chloride 5g/L, Sodium Lauryl Sulfate 0.1g/L | |
| Pail |