missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Lauryl Tryptose Broth
Specifications
Specifications
| Certifications/Compliance | CE, AOAC, BAM, CCAM, COMPF, ISO, SMD, SMWW |
| Content And Storage | Refrigerate |
| Form | Dehydrated |
| Media Type | Lauryl Tryptose Broth |
| Product Type | Nutrient Broth |
| Formulation | Tryptose 20g/L, Lactose 5g/L, Dipotassium Phosphate 2.75g/L, Monopotassium Phosphate 2.75g/L, Sodium Chloride 5g/L, Sodium Lauryl Sulfate 0.1g/L |
| For Use With (Application) | Detection of Coliform organisms in materials of sanitary importance |
| Packaging Type | Bottle |
| Quantity | 500 g |