missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Granulated Agar

Catalog No. p-4886723
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
10,000 g
2000 g
500 g
Catalog No. Quantity
DF0145070 2000 g
DF0145080 10,000 g
DF0145170 500 g
3 options

Catalog No. DF0145070

Supplier: BD 214510

Only null left

Used as a solidifying agent for culture media. BD Difco™ Dehydrated Culture Media: Granulated Agar is carefully monitored for cultural response, solubility and gelation temperature.

Applications:

  • Solidifying agent for culture media
  • Granulated

Specifications

Certifications/Compliance ISO/DIS, 11014
Concentration 1.5%
Additive Agar
Form Dehydrated Granule
For Use With (Application) General bacteriological purposes
Packaging Type Container
Quantity 2000 g