missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Granulated Agar

Catalog No. DF0145070
Change view
Click to view available options
Quantity:
10,000 g
500 g
2000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0145070 2000 g
DF0145080 10,000 g
DF0145170 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. DF0145070 Supplier BD Supplier No. 214510

Used as a solidifying agent for culture media. BD Difco™ Dehydrated Culture Media: Granulated Agar is carefully monitored for cultural response, solubility and gelation temperature.

Applications:

  • Solidifying agent for culture media
  • Granulated

Specifications

Certifications/Compliance ISO/DIS, 11014
Concentration 1.5%
Additive Agar
Form Dehydrated Granule
For Use With (Application) General bacteriological purposes
Packaging Type Container
Quantity 2000 g