missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Difco™ Beef Extract
GREENER_CHOICE

Catalog No. DF0126170
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
DF0126170 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. DF0126170 Supplier Gibco™ Supplier No. 212610

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Difco Beef Extract is an animal origin peptone derived from an infusion of beef. This nutritional supplement provides a mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins.

Difco Beef Extract is an animal origin peptone derived from an infusion of beef. This nutritional supplement provides a mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins. Provided as a paste, customers have used Difco Beef Extract in a variety of microbial fermentation cell lines, including Escherichia coli, Bacillus subtillis, Pasteurella multocida, and Streptococcus species to produce numerous animal health vaccines.

Features:

  • An animal origin nutritional supplement
  • A mixture of peptides, individual amino acids, nucleotides, organic acids, minerals, and vitamins

Applications of Difco Beef Extract include:

  • Use in a variety of microbial fermentation cell lines
  • Used to produce numerous animal health vaccines

Specifications

Certifications/Compliance LUO
Content And Storage Store at 2°C to 25°C in cool, dry conditions in well-sealed containers.
Form Paste
Product Type Bacterial Media
Format Bottle
For Use With (Application) Cultivation of heterotrophic microorganisms
Product Line Difco
Quantity 500 g
Source Animal origin-free