missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Culture Media Additives: Bovine Albumin 5%

To enrich media for cultivating and isolating large variety of microorganisms and tissue cells

Supplier:  BD 215330

Catalog No. B215330


Only null left
Explore available promotions

Description

Description

Bovine Albumin 5%

  • Used for isolating mycobacteria and enriching digested contaminated specimens

Specifications

Specifications

CE
5%
2°C to 8°C
Bovine Albumin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions