missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Adjuvants

Catalog No. DF0638607
Change view
Click to view available options
Product Type:
Complete H37Ra
Freund's Complete Adjuvant
Freund's Incomplete Adjuvant
Mycobacterium butyricum
Mycobacterium tuberculosis H37Ra
Quantity:
6 x 100 mg
6 x 10 mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
DF0638607 Freund's Complete Adjuvant 6 x 10 mL
DF0639606 Freund's Incomplete Adjuvant 6 x 10 mL
DF0640337 Mycobacterium butyricum 6 x 100 mg
DF3113605 Complete H37Ra 6 x 10 mL
DF3114338 Mycobacterium tuberculosis H37Ra 6 x 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Not intended for human or therapeutic use.
Catalog No. DF0638607 Supplier BD Supplier No. 263810

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Not intended for human or therapeutic use.

Used to prepare antigen-adjuvant emulsions. BD™ Difco™ Adjuvants are used in immunological studies with laboratory animals. (Not intended for human or therapeutic use.)

  • Adjuvants are mixed with aqueous antigen solution or suspension
  • Once injected, they provide a pool of emulsified antigen at the injection site and allow the antigen to be released over an extended period of time
  • With adjuvants, the immune response to antigens (especially soluble antigens) is enhanced and prolonged
  • Use Incomplete Freund adjuvant to avoid the use of acid-fast bacilli; however, the antibody response is not as marked as with the complete adjuvant

Specifications

Formulation Suspension of 5mg M. butyricum in a mixture of paraffin oil and an emulsifying agent
Product Type Freund's Complete Adjuvant
For Use With (Application) Immunological studies with laboratory animals
Quantity 6 x 10 mL
Content And Storage Store in cool, dry conditions in well sealed containers, away from oxidizing agents.