missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Sterile Finger Dab Plate: Trypticase Soy Agar w/Lecithin and Polysorbate 80
Plated media in gamma-irradiated packs for isolator systems multiwrap packaging protects media from vaporized hydrogen peroxide exposure
$963.08 - $963.08
Specifications
| Color | Light Beige |
|---|---|
| Format | 150 x 15mm Finger Dab |
| Manufacturing Origin | United States |
| Media Type | Trypticase™ Soy Agar with Lecithin and Polysorbate 80 |
| Sterility | Sterile |
Description
- Available in 100 x 15mm Style Plates
Specifications
| Light Beige | |
| United States | |
| Sterile |
| 150 x 15mm Finger Dab | |
| Trypticase™ Soy Agar with Lecithin and Polysorbate 80 | |
| Plate |