missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Sterile Finger Dab Plate: Trypticase Soy Agar w/Lecithin and Polysorbate 80
Description
- Available in 100 x 15mm Style Plates
Specifications
Specifications
| Certifications/Compliance | ISO/DIS, 11014 |
| Color | Light Beige |
| Format | 150 x 15mm Finger Dab |
| Manufacturing Origin | United States |
| Media Type | Trypticase™ Soy Agar with Lecithin and Polysorbate 80 |
| Sterility | Sterile |
| Packaging Type | Plate |
| Quantity | 100/Pk. |