Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Discs
Allow addition of antimicrobial agents to culture media.
$29.32 - $62.45
| Catalog Number | Mfr. No. | Concentration | Antimicrobial Agents | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Concentration | Antimicrobial Agents | Price | Quantity | ||||
|
B31575
|
BD
231575 |
25 μg | Ethambutol |
Each for $46.23
|
|
||||
|
B31570
|
BD
231570 |
50 μg | Streptomycin |
Each for $46.23
|
|
||||
|
B31571
|
BD
231571 |
1 μg | Isoniazid |
Each for $62.39
|
|
||||
|
B31572
|
BD
231572 |
5 μg | Isoniazid |
Each for $62.45
|
|
||||
|
B31576
|
BD
231576 |
50 μg | Ethambutol |
Each for $46.23
|
|
||||
|
B31577
|
BD
231577 |
25 μg | Ethionamide |
Each for $29.32
|
|
||||
|
B31578
|
BD
231578 |
25 μg | Rifampin |
Each for $46.23
|
|
||||
Specifications
| Disc | |
| Each |
Not for use in ordinary susceptibility plate tests.