Learn More
BD BBL™ Sensi-Disc™ Antimicrobial Discs
Allow addition of antimicrobial agents to culture media.
Supplier: BD 231576
Specifications
| Ethambutol | |
| 50 μg | |
| Disc | |
| Each |
Not for use in ordinary susceptibility plate tests.