missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Rabbit Coagulase Plasma
Qualitative determination of Staphylococci pathogenicity using direct tube method
$340.58 - $694.55
Specifications
| Certifications/Compliance | CE |
|---|---|
| Content And Storage | 2°C to 8°C |
| Form | Lyophilized |
| Product Type | Coagulase Rabbit Plasma |
| For Use With (Application) | Qualitative determination of Staphylococci pathogenicity using direct tube method |
Description
Coagulase binds plasma fibrinogen, causing the organisms to agglutinate or plasma to clot. BBL Coagulase Plasma, Rabbit and BBL Coagulase Plasma Rabbit with EDTA are recommended for performing the direct tube test. At the time of use, test both positive and negative control cultures to check performance of the coagulase plasma.
- Product type: diagnostic reagents and adjuvants
- Groupings: C Products
- Type: Coagulase Testing
- Shelf Life: 1095 days
- Label has Natural Rubber Latex statement
Certifications
CE marked
Specifications
| CE | |
| Lyophilized | |
| Qualitative determination of Staphylococci pathogenicity using direct tube method |
| 2°C to 8°C | |
| Coagulase Rabbit Plasma | |
| Rabbit |
Safety and Handling
ShelfLife : 1095 Days