missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Rabbit Coagulase Plasma

Catalog No. B40826
Change view
Click to view available options
Quantity:
10 x 3 mL
10 x 15 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40826 10 x 15 mL
B40827 10 x 3 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40826 Supplier BD Supplier No. 240826
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Qualitative determination of Staphylococci pathogenicity using direct tube method

Coagulase binds plasma fibrinogen, causing the organisms to agglutinate or plasma to clot. BBL Coagulase Plasma, Rabbit and BBL Coagulase Plasma Rabbit with EDTA are recommended for performing the direct tube test. At the time of use, test both positive and negative control cultures to check performance of the coagulase plasma.

  • Product type: diagnostic reagents and adjuvants
  • Groupings: C Products
  • Type: Coagulase Testing
  • Shelf Life: 1095 days
  • Label has Natural Rubber Latex statement

Certifications

CE marked

Specifications

Certifications/Compliance CE
Content And Storage 2°C to 8°C
Form Lyophilized
Product Type Coagulase Rabbit Plasma
For Use With (Application) Qualitative determination of Staphylococci pathogenicity using direct tube method
Quantity 10 x 15 mL
Species Rabbit