missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BBL™ Mycoflask™ Thioglycollate (Fluid) Prepared Media
Used for the sterility testing of biologics and for the cultivation of anaerobes, aerobes and microaerophiles
$361.20 - $549.21
Specifications
| Content And Storage | 2°C to 8°C |
|---|---|
| Form | Liquid |
| Manufacturing Origin | United States |
| Media Type | Thioglycollate (Fluid) |
| Formulation | Pancreatic digest of Casein 15g/L, L-Cystine 0.5g/L, Dextrose (anhydrous) 5g/L, Yeast extract 5g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Resazurin 0.001g/L, Agar 0.75g/L |
| Catalog Number | Mfr. No. | Quantity | Color | Media Type | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Color | Media Type | Price | Quantity | |||
|
B20889
|
BD
220889 |
100 x 20 mL | Beige | Thioglycollate (Fluid) |
Pack of 100 Each for $549.21
|
|
|||
|
B21196
|
BD
221196 |
100 x 8 mL | Light Beige | Thioglycollate (Fluid) |
Pack of 100 Each for $361.20
|
|
|||
Description
This medium is capable of supporting good growth of a great variety of fastidious organisms, of both pathogenic and nonpathogenic species.
- Neutralize antibacterial activity of mercurial compounds
- Before use ∽30% of media should oxidized
Tubes with tight caps should be opened carefully to avoid injury due to breakage of glass.
Specifications
| 2°C to 8°C | |
| United States | |
| Pancreatic digest of Casein 15g/L, L-Cystine 0.5g/L, Dextrose (anhydrous) 5g/L, Yeast extract 5g/L, Sodium Chloride 2.5g/L, Sodium Thioglycollate 0.5g/L, Resazurin 0.001g/L, Agar 0.75g/L | |
| Tube |
| Liquid | |
| Thioglycollate (Fluid) | |
| Sterility testing of biologics and for the cultivation of anaerobes, aerobes and microaerophiles |
For in vitro diagnostic use.